Assay Details
| Gene Symbol: LOC101717854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846414.3 | XP_004846471.1 | 5 - 6 | 914 | 71 | |
| XM_004846415.3 | XP_004846472.1 | 5 - 6 | 752 | 71 | ||
| XM_004846416.3 | XP_004846473.1 | 5 - 6 | 918 | 71 | ||
| XM_004846417.3 | XP_004846474.1 | 5 - 6 | 917 | 71 | ||
| XM_021259717.1 | XP_021115376.1 | 5 - 6 | 768 | 71 | ||
More Information
Set Membership: |
|

LOC101717854
Genomic Map