Assay Details
| Gene Symbol: Hnrnpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850548.3 | XP_004850605.1 | 9 - 10 | 1230 | 60 | |
| XM_013074407.2 | XP_012929861.1 | 10 - 11 | 1484 | 60 | ||
| XM_013074408.2 | XP_012929862.1 | 10 - 11 | 1501 | 60 | ||
| XM_013074409.2 | XP_012929863.1 | 9 - 10 | 1302 | 60 | ||
| XM_021263036.1 | XP_021118695.1 | 10 - 11 | 1511 | 60 | ||
More Information
Set Membership: |
|

Hnrnpr
Genomic Map