Assay Details
Gene Symbol: Dpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004858047.3 | XP_004858104.1 | 1 - 2 | 195 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
