Assay Details
| Gene Symbol: Cdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861357.3 | XP_004861414.1 | 1 - 2 | 303 | 70 | |
| XM_004861358.3 | XP_004861415.1 | 1 - 2 | 303 | 70 | ||
| XM_021241487.1 | XP_021097146.1 | 1 - 2 | 303 | 70 | ||
| XM_021241488.1 | XP_021097147.1 | 1 - 2 | 303 | 70 | ||
More Information
Set Membership: |
|

Cdk2
Genomic Map