Assay Details
| Gene Symbol: Ldb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847238.3 | XP_004847295.1 | 5 - 6 | 852 | 72 | |
| XM_004847239.3 | XP_004847296.1 | 5 - 6 | 858 | 72 | ||
| XM_004847240.3 | XP_004847297.1 | 5 - 6 | 856 | 72 | ||
| XM_004847242.3 | XP_004847299.1 | 5 - 6 | 858 | 72 | ||
| XM_004847243.3 | XP_004847300.1 | 5 - 6 | 858 | 72 | ||
| XM_004847246.3 | XP_004847303.1 | 5 - 6 | 855 | 72 | ||
| XM_004847247.3 | XP_004847304.1 | 5 - 6 | 855 | 72 | ||
| XM_013073082.2 | XP_012928536.1 | 5 - 6 | 863 | 72 | ||
| XM_013073083.2 | XP_012928537.1 | 5 - 6 | 858 | 72 | ||
| XM_021260246.1 | XP_021115905.1 | 5 - 6 | 860 | 72 | ||
| XM_021260247.1 | XP_021115906.1 | 5 - 6 | 858 | 72 | ||
| XM_021260248.1 | XP_021115907.1 | 5 - 6 | 863 | 72 | ||
More Information
Set Membership: |
|

Ldb2
Genomic Map