Assay Details
| Gene Symbol: Mcee | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852658.3 | XP_004852715.1 | 2 - 3 | 466 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcee| Gene Symbol: Mcee | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852658.3 | XP_004852715.1 | 2 - 3 | 466 | 91 | |
Set Membership: |
|