Assay Details
| Gene Symbol: Gsk3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835655.3 | XP_004835712.1 | 4 - 5 | 1497 | 81 | |
| XM_004835656.3 | XP_004835713.1 | 4 - 5 | 1489 | 81 | ||
| XM_004835658.3 | XP_004835715.1 | 4 - 5 | 531 | 81 | ||
| XM_021263075.1 | XP_021118734.1 | 4 - 5 | 460 | 81 | ||
| XM_021263083.1 | XP_021118742.1 | 4 - 5 | 1504 | 81 | ||
More Information
Set Membership: |
|

Gsk3b
Genomic Map