Assay Details
| Gene Symbol: LOC101704457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021262845.1 | XP_021118504.1 | 1 - 2 | 129 | 80 | |
| XM_021262846.1 | XP_021118505.1 | 9 - 10 | 1406 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021262845.1 transcripts.
Set Membership: |
|

LOC101704457
Genomic Map