Assay Details
| Gene Symbol: Zhx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837997.3 | XP_004838054.1 | 3 - 4 | 3110 | 98 | |
| XM_021252436.1 | XP_021108095.1 | 3 - 4 | 3101 | 98 | ||
More Information
Set Membership: |
|

Zhx1
Genomic Map