Assay Details
| Gene Symbol: Ccni | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848026.3 | XP_004848083.1 | 5 - 6 | 1018 | 145 | |
| XM_004848027.3 | XP_004848084.1 | 4 - 5 | 943 | 145 | ||
| XM_021261001.1 | XP_021116660.1 | 5 - 6 | 527 | 145 | ||
| XM_021261003.1 | XP_021116662.1 | 5 - 6 | 1019 | 145 | ||
| XM_021261004.1 | XP_021116663.1 | 5 - 6 | 865 | 145 | ||
More Information
Set Membership: |
|

Ccni
Genomic Map