Assay Details
| Gene Symbol: Wasf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854817.3 | XP_004854874.1 | 9 - 10 | 1606 | 64 | |
| XM_004854818.3 | XP_004854875.1 | 9 - 10 | 1597 | 64 | ||
More Information
Set Membership: |
|

Wasf3
Genomic Map