Assay Details
| Gene Symbol: Etv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845875.2 | XP_004845932.1 | 3 - 4 | 548 | 98 | |
| XM_021259174.1 | XP_021114833.1 | 3 - 4 | 546 | 98 | ||
| XM_021259175.1 | XP_021114834.1 | 3 - 4 | 551 | 98 | ||
| XM_021259176.1 | XP_021114835.1 | 2 - 3 | 417 | 98 | ||
More Information
Set Membership: |
|

Etv6
Genomic Map