Assay Details
| Gene Symbol: Cbfa2t2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867938.3 | XP_004867995.1 | 8 - 9 | 1388 | 65 | |
| XM_004867939.3 | XP_004867996.1 | 8 - 9 | 1388 | 65 | ||
| XM_021246781.1 | XP_021102440.1 | 8 - 9 | 1426 | 65 | ||
| XM_021246782.1 | XP_021102441.1 | 8 - 9 | 1426 | 65 | ||
| XM_021246783.1 | XP_021102442.1 | 8 - 9 | 1395 | 65 | ||
| XM_021246784.1 | XP_021102443.1 | 8 - 9 | 1257 | 65 | ||
| XM_021246785.1 | XP_021102444.1 | 7 - 8 | 1282 | 65 | ||
| XM_021246786.1 | XP_021102445.1 | 8 - 9 | 1427 | 65 | ||
More Information
Set Membership: |
|

Cbfa2t2
Genomic Map