Assay Details
| Gene Symbol: Tp53inp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850127.2 | XP_004850184.1 | 3 - 4 | 525 | 72 | |
| XM_004850129.2 | XP_004850186.1 | 3 - 4 | 549 | 72 | ||
| XM_004850131.3 | XP_004850188.1 | 3 - 4 | 525 | 72 | ||
More Information
Set Membership: |
|

Tp53inp1
Genomic Map