Assay Details
Gene Symbol: Wdr53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004834701.3 | XP_004834758.1 | 2 - 3 | 659 | 63 | |
XM_021255031.1 | XP_021110690.1 | 2 - 3 | 671 | 63 | ||
XM_021255036.1 | XP_021110695.1 | 2 - 3 | 1146 | 63 | ||
XM_021255045.1 | XP_021110704.1 | 3 - 4 | 468 | 63 | ||
XM_021255046.1 | XP_021110705.1 | 3 - 4 | 996 | 63 | ||
XM_021255052.1 | XP_021110711.1 | 3 - 4 | 468 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
