Assay Details
| Gene Symbol: Gadd45g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851310.2 | XP_004851367.1 | 3 - 4 | 487 | 52 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gadd45g| Gene Symbol: Gadd45g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851310.2 | XP_004851367.1 | 3 - 4 | 487 | 52 | |
Set Membership: |
|