Assay Details
| Gene Symbol: Cmtm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857826.2 | XP_004857883.1 | 3 - 4 | 544 | 71 | |
| XM_004857827.2 | XP_004857884.1 | 4 - 5 | 505 | 71 | ||
More Information
Set Membership: |
|

Cmtm8
Genomic Map