Assay Details
Gene Symbol: Syngr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004867015.2 | XP_004867072.1 | 3 - 4 | 540 | 60 | |
XM_004867016.2 | XP_004867073.1 | 3 - 4 | 493 | 60 | ||
XM_004867017.2 | XP_004867074.1 | 3 - 4 | 469 | 60 | ||
XM_021246261.1 | XP_021101920.1 | 3 - 4 | 417 | 60 | ||
XM_021246262.1 | XP_021101921.1 | 4 - 5 | 595 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
