Assay Details
| Gene Symbol: Txnl4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842920.3 | XP_004842977.1 | 3 - 4 | 634 | 73 | |
| XM_021256761.1 | XP_021112420.1 | 3 - 4 | 498 | 73 | ||
More Information
Set Membership: |
|

Txnl4b
Genomic Map