Assay Details
| Gene Symbol: Fosb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873189.2 | XP_004873246.1 | 3 - 4 | 559 | 64 | |
| XM_021250893.1 | XP_021106552.1 | 3 - 4 | 559 | 64 | ||
More Information
Set Membership: |
|

Fosb
Genomic Map