Assay Details
| Gene Symbol: Pcyt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872229.2 | XP_004872286.1 | 5 - 6 | 695 | 67 | |
| XM_004872230.2 | XP_004872287.1 | 6 - 7 | 782 | 67 | ||
| XM_004872232.2 | XP_004872289.1 | 6 - 7 | 776 | 67 | ||
| XM_004872233.2 | XP_004872290.1 | 4 - 5 | 578 | 67 | ||
| XM_013068315.2 | XP_012923769.1 | 6 - 7 | 780 | 67 | ||
| XM_021250183.1 | XP_021105842.1 | 6 - 7 | 895 | 67 | ||
| XM_021250184.1 | XP_021105843.1 | 5 - 6 | 665 | 67 | ||
More Information
Set Membership: |
|

Pcyt1b
Genomic Map