Assay Details
| Gene Symbol: Rgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862773.2 | XP_004862830.1 | 3 - 4 | 370 | 92 | |
| XM_021242924.1 | XP_021098583.1 | 2 - 3 | 289 | 92 | ||
More Information
Set Membership: |
|

Rgs1
Genomic Map