Assay Details
| Gene Symbol: Srsf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837314.3 | XP_004837371.1 | 6 - 7 | 746 | 76 | |
| XM_004837315.2 | XP_004837372.1 | 6 - 7 | 661 | 76 | ||
| XM_004837316.3 | XP_004837373.1 | 6 - 7 | 611 | 76 | ||
| XM_021247208.1 | XP_021102867.1 | 6 - 7 | 644 | 76 | ||
| XM_021247211.1 | XP_021102870.1 | 6 - 7 | 661 | 76 | ||
More Information
Set Membership: |
|

Srsf5
Genomic Map