Assay Details
| Gene Symbol: Jazf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839497.2 | XP_004839554.1 | 1 - 2 | 225 | 75 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jazf1| Gene Symbol: Jazf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839497.2 | XP_004839554.1 | 1 - 2 | 225 | 75 | |