Assay Details
| Gene Symbol: Elovl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867583.3 | XP_004867640.1 | 3 - 4 | 409 | 77 | |
| XM_004867584.3 | XP_004867641.1 | 3 - 4 | 449 | 77 | ||
| XM_021246587.1 | XP_021102246.1 | 2 - 3 | 224 | 77 | ||
| XM_021246588.1 | XP_021102247.1 | 2 - 3 | 301 | 77 | ||
More Information
Set Membership: |
|

Elovl6
Genomic Map