Assay Details
| Gene Symbol: Colec12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853176.2 | XP_004853233.1 | 3 - 4 | 312 | 69 | |
| XM_021265180.1 | XP_021120839.1 | 3 - 4 | 361 | 69 | ||
| XM_021265182.1 | XP_021120841.1 | 2 - 3 | 184 | 69 | ||
More Information
Set Membership: |
|

Colec12
Genomic Map