Assay Details
| Gene Symbol: Elf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844262.3 | XP_004844319.1 | 8 - 9 | 1624 | 88 | |
| XM_004844263.3 | XP_004844320.1 | 8 - 9 | 1552 | 88 | ||
| XM_021257914.1 | XP_021113573.1 | 8 - 9 | 1698 | 88 | ||
| XM_021257915.1 | XP_021113574.1 | 9 - 10 | 1731 | 88 | ||
More Information
Set Membership: |
|

Elf1
Genomic Map