Assay Details
| Gene Symbol: Frmd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835541.3 | XP_004835598.1 | 13 - 14 | 1850 | 63 | |
| XM_004835542.3 | XP_004835599.1 | 13 - 14 | 1826 | 63 | ||
| XM_021262274.1 | XP_021117933.1 | 13 - 14 | 1801 | 63 | ||
| XM_021262279.1 | XP_021117938.1 | 13 - 14 | 1746 | 63 | ||
| XM_021262285.1 | XP_021117944.1 | 13 - 14 | 1793 | 63 | ||
| XM_021262294.1 | XP_021117953.1 | 12 - 13 | 1514 | 63 | ||
More Information
Set Membership: |
|

Frmd6
Genomic Map