Assay Details
| Gene Symbol: Ppil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846674.3 | XP_004846731.1 | 2 - 3 | 473 | 101 | |
| XM_004846675.2 | XP_004846732.1 | 2 - 3 | 425 | 101 | ||
More Information
Set Membership: |
|

Ppil1
Genomic Map