Assay Details
| Gene Symbol: Psmc3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859324.3 | XP_004859381.1 | 3 - 4 | 398 | 58 | |
| XM_021240282.1 | XP_021095941.1 | 3 - 4 | 1149 | 58 | ||
More Information
Set Membership: |
|

Psmc3ip
Genomic Map