Assay Details
| Gene Symbol: Ckmt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861701.3 | XP_004861758.1 | 8 - 9 | 1056 | 68 | |
| XM_021241710.1 | XP_021097369.1 | 8 - 9 | 1059 | 68 | ||
More Information
Set Membership: |
|

Ckmt1a
Genomic Map