Assay Details
Gene Symbol: Immp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021253897.1 | XP_021109556.1 | 72 | 63 | ||
XM_021253898.1 | XP_021109557.1 | 91 | 63 | |||
XM_021253899.1 | XP_021109558.1 | 135 | 63 | |||
XM_021253900.1 | XP_021109559.1 | 1 - 2 | 72 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021253897.1, XM_021253899.1, XM_021253900.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
