Assay Details
| Gene Symbol: Ubxn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834698.2 | XP_004834755.1 | 9 - 10 | 1279 | 76 | |
| XM_013071927.2 | XP_012927381.1 | 9 - 10 | 1236 | 76 | ||
More Information
Set Membership: |
|

Ubxn7
Genomic Map