Assay Details
| Gene Symbol: Rnf167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857304.2 | XP_004857361.1 | 6 - 7 | 1162 | 63 | |
| XM_004857305.2 | XP_004857362.1 | 6 - 7 | 1110 | 63 | ||
| XM_013076887.2 | XP_012932341.1 | 6 - 7 | 1149 | 63 | ||
More Information
Set Membership: |
|

Rnf167
Genomic Map