Assay Details
| Gene Symbol: Rdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842052.3 | XP_004842109.1 | 3 - 4 | 742 | 84 | |
| XM_021256019.1 | XP_021111678.1 | 3 - 4 | 742 | 84 | ||
More Information
Set Membership: |
|

Rdh10
Genomic Map