Assay Details
| Gene Symbol: Mat2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836775.3 | XP_004836832.2 | 4 - 5 | 651 | 70 | |
| XM_021240011.1 | XP_021095670.1 | 658 | 70 | |||
More Information
Set Membership: |
|

Mat2b
Genomic Map