Assay Details
| Gene Symbol: LOC101718541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845458.3 | XP_004845515.1 | 1 - 2 | 371 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004845458.3 transcripts.
Set Membership: |
|

LOC101718541
Genomic Map