Assay Details
| Gene Symbol: Klf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859884.2 | XP_004859941.1 | 3 - 4 | 933 | 85 | |
| XM_021240394.1 | XP_021096053.1 | 3 - 4 | 952 | 85 | ||
More Information
Set Membership: |
|

Klf6
Genomic Map