Assay Details
| Gene Symbol: LOC101710326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848950.3 | XP_004849007.1 | 6 - 7 | 668 | 73 | |
| XM_004848951.3 | XP_004849008.1 | 6 - 7 | 646 | 73 | ||
| XM_021261737.1 | XP_021117396.1 | 6 - 7 | 669 | 73 | ||
More Information
Set Membership: |
|

LOC101710326
Genomic Map