Assay Details
| Gene Symbol: Sqor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835467.3 | XP_004835524.1 | 5 - 6 | 772 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sqor| Gene Symbol: Sqor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835467.3 | XP_004835524.1 | 5 - 6 | 772 | 77 | |
Set Membership: |
|