Assay Details
| Gene Symbol: Mrps10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846574.3 | XP_004846631.1 | 6 - 7 | 570 | 65 | |
| XM_004846575.3 | XP_004846632.1 | 6 - 7 | 567 | 65 | ||
| XM_021259816.1 | XP_021115475.1 | 7 - 8 | 591 | 65 | ||
| XM_021259817.1 | XP_021115476.1 | 7 - 8 | 588 | 65 | ||
More Information
Set Membership: |
|

Mrps10
Genomic Map