Assay Details
| Gene Symbol: Scara3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848395.2 | XP_004848452.1 | 5 - 6 | 1612 | 67 | |
| XM_004848396.2 | XP_004848453.1 | 4 - 5 | 1513 | 67 | ||
| XM_013073474.2 | XP_012928928.1 | 5 - 6 | 1604 | 67 | ||
| XM_021261320.1 | XP_021116979.1 | 4 - 5 | 1514 | 67 | ||
More Information
Set Membership: |
|

Scara3
Genomic Map