Assay Details
| Gene Symbol: Ppm1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861132.3 | XP_004861189.1 | 3 - 4 | 1108 | 77 | |
| XM_004861133.3 | XP_004861190.1 | 3 - 4 | 615 | 77 | ||
| XM_021241527.1 | XP_021097186.1 | 4 - 5 | 1228 | 77 | ||
| XM_021241528.1 | XP_021097187.1 | 4 - 5 | 1235 | 77 | ||
| XM_021241529.1 | XP_021097188.1 | 4 - 5 | 1234 | 77 | ||
More Information
Set Membership: |
|

Ppm1h
Genomic Map