Assay Details
| Gene Symbol: Tgfb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837154.3 | XP_004837211.1 | 5 - 6 | 2237 | 53 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tgfb3| Gene Symbol: Tgfb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837154.3 | XP_004837211.1 | 5 - 6 | 2237 | 53 | |
Set Membership: |
|