Assay Details
| Gene Symbol: Adora1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862195.3 | XP_004862252.1 | 2 - 3 | 488 | 89 | |
| XM_004862196.3 | XP_004862253.1 | 2 - 3 | 484 | 89 | ||
| XM_004862197.3 | XP_004862254.1 | 1 - 2 | 562 | 89 | ||
More Information
Set Membership: |
|

Adora1
Genomic Map