Assay Details
| Gene Symbol: Nuak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862108.3 | XP_004862165.1 | 2 - 3 | 539 | 60 | |
| XM_004862110.2 | XP_004862167.1 | 3 - 4 | 348 | 60 | ||
More Information
Set Membership: |
|

Nuak2
Genomic Map