Assay Details
Gene Symbol: Sar1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004836557.2 | XP_004836614.1 | 6 - 7 | 635 | 69 | |
XM_004836558.2 | XP_004836615.1 | 7 - 8 | 731 | 69 | ||
XM_021241040.1 | XP_021096699.1 | 6 - 7 | 515 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
