Assay Details
| Gene Symbol: Lif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843430.3 | XP_004843487.1 | 1 - 2 | 237 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004843430.3 transcripts.
Set Membership: |
|

Lif
Genomic Map