Assay Details
| Gene Symbol: Lif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843430.3 | XP_004843487.1 | 2 - 3 | 415 | 80 | |
| XM_021257192.1 | XP_021112851.1 | 2 - 3 | 697 | 80 | ||
More Information
Set Membership: |
|

Lif
Genomic Map