Assay Details
| Gene Symbol: Spx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845777.2 | XP_004845834.1 | 4 - 5 | 2668 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spx| Gene Symbol: Spx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845777.2 | XP_004845834.1 | 4 - 5 | 2668 | 82 | |
Set Membership: |
|